missing translation for 'onlineSavingsMsg'
Learn More

Quercetin dihydrate, 97%

Catalog No. AAA1580736
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1580736 500 g
AAA1580714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1580736 Supplier Thermo Scientific Chemicals Supplier No. A1580736
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 6151-25-3 | C15H14O9 | 338.27 g/mol

Quercetin dihydrate used as a Inhibitor of protein tyrosine kinase, mitochondrial ATPase and phosphodiesterase inhibitor. It Inhibits PI3-kinase activity and slightly inhibits PIP kinase activity.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Quercetin dihydrate used as a Inhibitor of protein tyrosine kinase, mitochondrial ATPase and phosphodiesterase inhibitor. It Inhibits PI3-kinase activity and slightly inhibits PIP kinase activity.

Solubility
Soluble in DMSO (68 mg/ml at 25°C), Acetic Acid (1 mg/ml), Aqueous Alkali (1 mg/ml), water (<1 at 25°C), and ethanol (21 mg/ml at 25°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 6151-25-3
Molecular Formula C15H14O9
Molecular Weight (g/mol) 338.27
MDL Number MFCD00149487
InChI Key GMGIWEZSKCNYSW-UHFFFAOYSA-N
Synonym quercetin dihydrate, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-chromen-4-one dihydrate, quercetine dihydrate, quercetin, dihydrate, ccris 3304, 3,3',4',5,7-pentahydroxyflavone dihydrate, quercetin dihydrate sophoretin, flavone, 3,3',4',5,7-pentahydroxy-, dihydrate, dsstox_cid_1219, 2-3,4-dihydroxyphenyl-3,5,7-trihydroxy-4h-1-benzopyran-4-one dihydrate
PubChem CID 5284452
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,5,7-trihydroxychromen-4-one;dihydrate
SMILES O.O.OC1=CC(O)=C2C(OC(=C(O)C2=O)C2=CC=C(O)C(O)=C2)=C1

Specifications

Melting Point 310°C to 315°C
Odor Odorless
Assay Percent Range 97%
Quantity 500 g
UN Number UN2811
Beilstein 317313
Merck Index 14,8034
Solubility Information Soluble in DMSO (68mg/ml at 25°C),Acetic Acid (1mg/ml),Aqueous Alkali (1mg/ml),water (<1 at 25°C),and ethanol (21mg/ml at 25°C).
Formula Weight 338.28 (302.25 Anhydrous)
Percent Purity 97%
Chemical Name or Material Quercetin dihydrate
Show More Show Less
Hazard Category H301-H341
Hazard Statement GHS H Statement
H301-H341
Toxic if swallowed.
Suspected of causing genetic defects.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P310-P308+P313-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 204-187-1
RTECSNumber LK8950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only