missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ QuEChERS Original 2003 Method Extraction Kits

Catalog No. p-7222020
Change view
Click to view available options
Sample Size:
10 g sample
Includes:
Salts in Pouches
Salts in Pouches, 50 mL Tubes
Salts in Pouches, 50 mL Tubes, Ceramic Homogenizers
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sample Size Includes
03254141 10 g sample Salts in Pouches, 50 mL Tubes, Ceramic Homogenizers
03254142 10 g sample Salts in Pouches, 50 mL Tubes
03254143 10 g sample Salts in Pouches
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 03254141 Supplier Thermo Scientific™ Supplier No. S110ORIGCHKIT
Only null left

Use Thermo Scientific™ QuEChERS Original 2003 Method Extraction Kits to analyze fruits and vegetables for pesticides, veterinary drug residues, and toxins.

Use Thermo Scientific™ QuEChERS Extraction Kits to prepare complex sample matrices such as fruits and vegetables for LC-MS or GC-MS analysis for pesticide residues, veterinary drug residues, and toxins using the Original 2003 Method. Our QuEChERS (Quick, Easy, Cheap, Effective, Rugged and Safe) kits contain pre-packaged, ready-weighed salts, sorbents, and buffers designed to streamline your workflow, provide excellent recoveries for a wide variety of analytes, minimize the potential for error, save time and money, and deliver high-quality, reproducible results.

Thermo Scientific™ QuEChERS Original 2003 Method Extraction Kits make sample preparation straight-forward and efficient. With our QuEChERS kits, you can confidently perform extractions and analyze fruit and vegetable samples for pesticide residues and other contaminants.

Three different extraction kit types are available to support the Original 2003 QuEChERS Method.

  • Extraction Salts in Pouches, no Tubes – for 10 g or 15 g samples
  • Extraction Salts in Pouches with 50 mL Tubes – for 10 g or 15 g samples
  • Extraction Salts in Pouches with 50 mL Tubes and Ceramic Homogenizers – for 10 g or 15 g samples

What is the purpose of the chemicals used in the Original 2003 Method?

  • Acetonitrile: used to perform single-phase extraction on a sample
  • Anhydrous MgSO4: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning step; removes remaining water from sample during dispersive solid phase extraction (dSPE) step
  • NaCl: induces phase separation between water content in sample and acetonitrile layer during liquid-liquid partitioning step; removes residual water from sample; reduces polar interference and enhances extraction
  • PSA: primary secondary amine, used to remove polar matrix components, citric co-extractives, and free fatty acids, from sample during dispersive solid phase extraction (dSPE) step

Specifications

Description QuEChERS Original Extraction Kit for 10 g sample; 4 g MgSO4, 1 g NaCl in Pouches; 50 mL Tubes; Ceramic Homogenizers; 50/Pk.
Type QuEChERS Extraction Kit
Includes Salts in Pouches, 50 mL Tubes, Ceramic Homogenizers
Method QuEChERS sample preparation for Original 2003 Method
Salt Composition 4 g MgSO4, 1 g NaCl
Sample Size 10 g sample
Sample Type All
Tube Size 50 mL