Learn More
Invitrogen™ Qubit™ Flex Starter Packages
Measure fluorescence simultaneously, and reduce assay variability, with the Qubit Flex NGS and Flex Quantitation Starter Kits, which offer highly accurate quantification of DNA, RNA, microRNA, and protein using the Qubit Flex Fluorometer .
Supplier: Invitrogen™ Q45894
Description
Invitrogen Qubit Flex starter kits combine a Qubit Flex Fluorometer with the reagents, plastics, accessories, and training necessary to get starting using the Qubit Flex Fluorometer for quantitation, next-generation sequencing, and endotoxin detection.
The Qubit Flex Fluorometer is a benchtop fluorometer designed for highly accurate quantification of DNA, RNA, microRNA, protein, and endotoxin. With the Qubit Flex Fluorometer you have the flexibility to directly measure fluorescence of up to eight samples simultaneously, which reduces assay variability. Paired with Qubit reagents, the Qubit Flex Fluorometer is part of an optimized workflow generating highly accurate and reproducible results.
Qubit Flex Quantitation Starter Kit (Cat. No. Q45894) includes:
• Qubit Flex Fluorometer (Cat. No. Q33326)
• Qubit Flex Assay Tube Strips, 125 strips (Cat. No. Q33252)
• Qubit 1X dsDNA HS Assay Kit, 100 assays (Cat. No. Q33230)
• Qubit RNA HS Assay Kit, 100 assays (Cat. No. Q32852)
• Qubit RNA BR Assay Kit, 100 assays (Cat. No. Q10210)
• Qubit Protein Assay Kit, 100 assays (Cat. No. Q33211)
• Qubit Flex System Verification Assay Kit (Cat. No. Q33254)
• Qubit Flex Reservoirs, 100 reservoirs (Cat. No. Q33253)
• WiFi Dongle (Cat. No. A26774)
• Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument
• Three-year Extended Warranty—if your instrument needs repair we will send you a factory-certified refurbished replacement
Qubit Flex NGS (Next-Generation Sequencing) Starter Kit (Cat. No. Q45893) includes:
• Qubit Flex Fluorometer (Cat. No. Q33326)
• Qubit Flex Assay Tubes Strips, 125 strips (Cat. No. Q33252)
• Qubit 1X dsDNA HS Assay Kit, 500 assays (Cat. No. Q33231)
• Qubit Flex Assay Reservoirs, 100 reservoirs (Cat. No. Q33253)
• WiFi Dongle (Cat. No. A26774)
• Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument
• Three-year Extended Warranty—if your instrument needs repair we will send you a factory-certified refurbished replacement
Qubit Flex Endotoxin Detection Starter Kit (Cat. No. Q32894) includes:
• Qubit Flex Fluorometer (Cat. No. Q33327)
• Qubit Flex Pyrogen Free Assay Tubes Strips, 120 strips (Cat. No. Q32893)
• Qubit Endotoxin Detection Assay Kit, 80 assays (Cat. No. Q32891)
• Qubit Flex System Verification Assay Kit (Cat. No. Q33254)
• WiFi Dongle (Cat. No. A26774)
• Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument
• Three-year Extended Warranty—if your instrument needs repair we will send you a factory-certified refurbished replacement
Specifications
1 to 20 ÎĽL | |
460 to 480 nm (Blue LED), 620 to 640 nm (Red LED) | |
Quantitation Starter Kit | |
1 kit |
Qubit™ Flex Fluorometer, Qubit™ Flex Power Cord, Wi-Fi Dongle, Qubit™ Flex Assay Tube Strips, and Qubit™ Assay Kits | |
Qubit | |
DNA Quantification, RNA Quantification, MicroRNA Quantification, Protein Quantification |
Promotions
-
Thermo Fisher Scientific
,
Invitrogen
Save Up to 14% with Fluorometer Starter Kits
Get combined savings of up to 14% when you buy an Invitrogen Qubit Fluorometer Kit.Expires: 12/31/25