Learn More
Invitrogen™ Qubit™ Flex Endotoxin Detection Starter Kit with Warranty
Description
Qubit Flex Endotoxin Detection Starter Kit contents
- Qubit Flex Fluorometer (Cat. No. Q33327)
- Qubit Flex Pyrogen Free Assay Tubes Strips, 120 strips (Cat. No. Q32893)
- Qubit Endotoxin Detection Assay Kit, 80 assays (Cat. No. Q32891)
- Qubit Flex System Verification Assay Kit (Cat. No. Q33254)
- WiFi Dongle (Cat. No. A26774)
- Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument
Qubit Flex Fluorometer
The Qubit Flex Fluorometer is designed for highly accurate and sensitive quantification of DNA, RNA, microRNA, protein, and endotoxin. This benchtop fluorometer offers the flexibility to directly measure fluorescence of up to eight samples simultaneously, which reduces assay variability. Paired with Qubit reagents, the Qubit Flex Fluorometer is part of an optimized workflow generating highly accurate and reproducible results. The fluorometer has a large screen and intuitive user interface, making it easy to select and run your desired assay with results displayed in just a few seconds.
Features of the Qubit Flex Fluorometer include:
- Improved throughput—measure multiple samples and quantify DNA, RNA, protein, and endotoxin up to 50% faster than single-sample readers
- Flexibility—measure up to eight samples simultaneously using 1–20 μL of sample
- Complete optimized system—works seamlessly with Qubit reagents to generate reliable, sensitive, and specific results
- On-board calculators—quickly generates Qubit working solution preparation instructions and determines the molarity of your samples
- Easy to use—a large 8-inch, state-of-the-art color touchscreen and intuitive user interface results in easy workflow navigation
- Data export flexibility—data is transferred easily as a CSV or PDF using the included Wi-Fi dongle, a USB drive, or connection to your network drive
- Several languages available—choices include English, French, Spanish, Italian, German, simplified Chinese, and Japanese
- Verification test—a pre-loaded program is included to verify performance of the instrument
- Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument
Comprehensive warranty options
The Qubit Flex Fluorometer comes with a standard 1-year warranty that covers repair costs including shipping, parts and labor. Service plans can be bundled with new instrument purchases to provide a 3-year warranty. The Starter Kit is offered with a choice of service plans:
- Three-year Rapid Exchange (REX) Warranty—replacement shipped out in ~1 business day for labs with critical timelines
- Three-year AB Repair Center (ABRC) Warranty—instrument repaired and returned in ~3 weeks, ideal for regulated labs required to maintain asset tagging consistency
21 CFR Part 11 Software Module
The Qubit Flex Security Software supports compliance with 21 CFR Part 11 FDA guidelines for security, auditing, and electronic (SAE) signatures using the SAE administration console. The 21 CFR Part 11 software can be purchased separately (Cat. No. Q31994).
Qubit Endotoxin Detection Assay Kit
The Qubit Endotoxin Detection Assay Kit is a sensitive and easy-to-use fluorescent endpoint assay for use on the Qubit Flex Fluorometer that uses amebocyte lysates to quantify endotoxin in protein, peptide, antibody, or nucleic acid samples. Amebocyte lysates are widely used as a simple and sensitive assay for the detection of endotoxin lipopolysaccharide (LPS), the membrane component of gram-negative bacteria.
Features of the Qubit Endotoxin Detection Assay Kit include:
- Highly sensitive with a broad range—detect as little as 0.01 EU/mL to 10.0 EU/mL
- Flexible—suitable for wide range of samples, including DNA, RNA, plasmids, antibodies, and more
- Easy to use—when paired with the Qubit Flex Fluorometer, calculations are performed automatically, helping reduce the potential for error
Specifications
Specifications
| Content And Storage | Qubit Flex Endotoxin Detection Starter Kits include: •Qubit Flex Fluorometer (Cat. No. Q33327) •Qubit Flex Pyrogen Free Assay Tubes Strips, 120 strips (Cat. No. Q32893) •Qubit Endotoxin Detection Assay Kit, 80 assays (Cat. No. Q32891) •Qubit Flex System Verification Assay Kit (Cat. No. Q33254) •WiFi Dongle (Cat. No. A26774) •Qubit Flex SmartStart Digital Educational Experience—includes interactive visual demonstrations of how to install, operate, and maintain your instrument |
| Detection Method | Fluorescence |
| Excitation Wavelength | 460 to 480 nm (Blue LED), 620 to 640 nm (Red LED) |
| For Use With (Equipment) | Qubit Flex Fluorometer |
| Product Line | Qubit |
| Type | Benchtop Fluorometer |
| For Use With (Application) | DNA, RNA, Protein and Endotoxin Quantification |
| Quantity | 1 Kit |
| Warranty | Standard Warranty |
For Research Use Only. Not for use in diagnostic procedures.