Learn More
Thermo Scientific Chemicals Quartz tubing, fused, 12mm (0.472in) OD, 8mm (0.315in) ID
Manufactured by fusing naturally occurring crystalline silica
Supplier: Thermo Scientific Chemicals 042274NT
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| Fused Quartz | |
| Fused Quartz Tubing |
Specifications
| Fused Quartz | |
| 25 mm | |
| Fused Quartz Tubing |
| 0.984 in. | |
| 122 cm |
RUO – Research Use Only