missing translation for 'onlineSavingsMsg'
Learn More

Quartz rod, fused, 5.0mm (0.197in) dia

CAS: 14808-60-7 | O2Si | 60.08 g/mol

Supplier:  Thermo Scientific Chemicals 042293NT

 View more versions of this product

Catalog No. AA42293NT


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2230°C
1710°C
Rod
O2Si
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
O=[Si]=O
60.08
CHEBI:30563
Quartz rod
14808-60-7
48 in.
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
silanedione
24261
Tube

Chemical Identifiers

14808-60-7
60.08
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
silanedione
O2Si
MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
CHEBI:30563
O=[Si]=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

P201-P202-P260-P281-P308+P313-P501c

H350-H373

EINECSNumber : 238-878-4

RTECSNumber : VV7330000

TSCA : U

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only