missing translation for 'onlineSavingsMsg'
Learn More

Promega Quantus™ Fluorometer

Catalog No. PRE6150
Encompass
Change view
Click to view available options
For Use With (Application):
Quantifying nucleic acids and proteins
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Application)
PRE6150 Quantifying nucleic acids and proteins
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRE6150 Supplier Promega Supplier No. E6150

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Quantus Fluorometer, USB Power Supply, USB Cord, Outlet Adapter Kit (4 adapters supplied for international compatibility), Quick Reference Guide, Operating Manual, Product Registration Card

The Quantus Fluorometer is a dual-channel fluorometer for your personal quantitation workflow. Designed to provide highly sensitive fluorescent detection when quantifying nucleic acids, the compact instrument is simple to operate.

  • Simple, Sensitive DNA and RNA Quantitation
  • Easy, intuitive controls
  • Ready to use with QuantiFluor(R) Dyes
  • Recommended for Next-Generation Sequencing

Recommended Storage

Store at 4° to 40°C under noncondensing conditions

Specifications

Description Store at 15 to 30°C
Calibration Type Single point
Detector Type Solid state silicon sensor
Includes 1/ea; Quantus Fluorometer, USB Power Supply, USB Cord, Outlet Adapter Kit (4 adapters supplied for international compatibility), Quick Reference Guide, Operating Manual, Product Registration Card
Excitation Wavelength 495 nm (Blue), 640 nm (Red)
Dynamic Range Five orders of magnitude, assay-dependent
Emission Wavelength Red: 660 to 720nm, Blue: 510 to 580nm
For Use With (Application) Quantifying nucleic acids and proteins
Software Quantus software
Dimensions (L x W x H) 22.7 × 11.5 × 4.5cm
Weight (Metric) 400 g
Voltage 5 V
Amperage 0.2 A max.
Show More Show Less