missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 7 Pro Dx Block Upgrade Kit, 96-well, IVD

Catalog No. A51231
Change view
Click to view available options
Quantity:
1 kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A51231 1 kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A51231 Supplier Applied Biosystems™ Supplier No. A51231

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The IVD QuantStudio™ 7 Pro Dx Block Upgrade Kit is an optional accessory for the QuantStudio 7 Pro Dx Real-Time qPCR System.

  • Kit is an optional accessory for the QuantStudio™ 7 Pro Dx Real-Time qPCR System
  • Interchangeable blocks enable application versatility with easy, powered-up interchange of block types
  • The QuantStudio 7 Pro Dx system accommodates the interchange of 96-well (0.2 mL) and 384-well blocks
  • System is delivered with one block type; other block types may be purchased separately

Specifications

Content And Storage IVD 96-well, 0.2 mL block, IVD 96-well, 0.2 mL heated cover, and accessories
For Use With (Equipment) QuantStudio™ 7 Pro Dx
Product Line QuantStudio
Product Type Upgrade Kit
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Room Temperature
Quantity 1 kit

For In Vitro Diagnostic Use.