missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ QuantStudio™ 7 Pro Dx Block Upgrade Kit, 384-well, IVD
Description
- Kit is an optional accessory for the QuantStudio™ 7 Pro Dx Real-Time qPCR System
- Interchangeable blocks enable application versatility with easy, powered-up interchange of block types
- The QuantStudio 7 Pro Dx system accommodates the interchange of 96-well (0.2 mL) and 384-well blocks
- System is delivered with one block type; other block types may be purchased separately
Specifications
Specifications
| Content And Storage | IVD 384-well block, IVD 384-well heated cover, and accessories |
| For Use With (Equipment) | QuantStudio™ 7 Pro Dx |
| Product Line | QuantStudio |
| Product Type | Upgrade Kit |
| For Use With (Application) | Real Time PCR (qPCR) |
| Shipping Condition | Room Temperature |
| Quantity | 1 kit |
For In Vitro Diagnostic Use.