missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 5 Spectral Calibration Plate 1 (FAM™, VIC™, ROX™, TAMRA™, and SYBR™ dyes), 384-well

Catalog No. A26334
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26334 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A26334 Supplier Applied Biosystems™ Supplier No. A26334
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This plate establishes the pure dye spectra and multicomponent values for FAM™, VIC™, ROX™, TAMRA™, and SYBR™ dyes on Applied Biosystems™ QuantStudio™ 5 384-well real-time PCR system.

This plate establishes the pure dye spectra and multicomponent values for FAM™, VIC™, ROX™, TAMRA™, and SYBR™ dyes on Applied Biosystems™ QuantStudio™ 5 384-well real-time PCR system. It enables simultaneous multiple dye calibrations in one plate, significantly reducing effort and time required for dye calibration. Please refer to the instrument user guide for full instructions on usage.

Specifications

Format 384-well
Content And Storage Store at –20°C ± 5°C.

Calibrated Dye FAM, ROX, TAMRA™, VIC, SYBR™ Green
For Use With (Equipment) QuantStudio™ 5, QuantStudio™ 6 Pro, QuantStudio™ 7 Pro, QuantStudio™ 6 Flex (Software version 1.6 or later), QuantStudio™ 7 Flex (Software version 1.6 or later)
Product Type Calibration Plate
For Use With (Application) Spectral Calibration
Shipping Condition Dry Ice
Quantity 1 plate
Plate Compatibility 384-well
Calibration Type Spectral Calibration
PCR Method qPCR
Show More Show Less
Do I need to perform a normalization calibration for QuantStudio 6 and 7 Flex Real-Time PCR System Software 1.7 after running a QuantStudio 5 Spectral Calibration Plate 1, 384 well (Cat. No. A26334)?

No, you do not need to perform the normalization calibration. The following link, Real-Time PCR Instrument Calibration & Verification Kits, in the “QuantStudio 7 Flex” section, shows that normalization plates are not included if the software is version 1.6 or later.

Is the QuantStudio 5 Spectral Calibration Plate 1, 384 well (Cat. No. A26334) compatible with the QuantStudio 7 Flex Real-Time PCR System, 384 well (Cat. No. 4485701)?

Yes, the QuantStudio 5 Spectral Calibration Plate 1, 384 well is compatible with the QuantStudio 7 Flex Real-Time PCR System, 384 well, with version 1.6, or later, of the corresponding software. However, this plate is not compatible with previous versions of the software.

For Research Use Only. Not for use in diagnostic procedures.