missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 3D Digital PCR Chip Lids and Blades

Catalog No. 4485510
Encompass
Catalog No. 4485510 Supplier Applied Biosystems™ Supplier No. 4485510

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Needed to securely cover and seal the QuantStudio™ 3D Digital PCR Chip after reagent addition.

Simply load the reaction mix onto the chip using a blade, cover the chip with a chip lid, and seal the chip. The chip is then ready for amplification and analysis.

Specifications

Content And Storage 12 Chip Lids, 12 Loading Blades
For Use With (Equipment) GeneAmp™ 9700
Product Type Lids, Blades
For Use With (Application) Digital PCR, Real Time PCR (qPCR)
Description 12 Chips
Quantity 12 Chips