missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 3/5 Spectral Calibration Plate 3 (TAMRA™, NED™, Cy™5 dyes), 96-well, 0.1 mL

Catalog No. A26340
Change view
Click to view available options
Quantity:
1 Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26340 1 Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A26340 Supplier Applied Biosystems™ Supplier No. A26340
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

This plate establishes the pure dye spectra and multicomponent values for TAMRA™, NED™, and Cy™5 dyes on Applied Biosystems™ QuantStudio™ 3 and 5 96-well 0.1-mL real-time PCR systems.

This plate establishes the pure dye spectra and multicomponent values for TAMRA™, NED™, and Cy™5 dyes on Applied Biosystems™ QuantStudio™ 3 and 5 96-well 0.1-mL real-time PCR systems. It enables simultaneous multiple dye calibrations in one plate, significantly reducing effort and time required for dye calibration. Please refer to the instrument user guide for full instructions on usage.

Specifications

Format 96-well, 0.1 mL
Content And Storage Store at –20°C ± 5°C.

Calibrated Dye Cy™5, NED, TAMRA™
For Use With (Equipment) QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Pro, QuantStudio™ 7 Pro, QuantStudio™ 6 Flex (Software version 1.6 or later), QuantStudio™ 7 Flex (Software version 1.6 or later)
Product Line Cy5, NED, QuantStudio, TAMRA
Product Type Calibration Plate
For Use With (Application) Spectral Calibration
Shipping Condition Dry Ice
Quantity 1 Plate
Plate Compatibility 96-well, 0.1 mL
Calibration Type Spectral Calibration
PCR Method qPCR
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.