missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 3/5 Spectral Calibration Plate 1 (FAM™, VIC™, ROX™, SYBR™ dyes), 96-well, 0.1 mL

Catalog No. A26336
Change view
Click to view available options
Quantity:
1 plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A26336 1 plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A26336 Supplier Applied Biosystems™ Supplier No. A26336
Get Up to $3,000 of qPCR Products with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Plate establishes the pure dye spectra and multicomponent values for FAM™, VIC™, ROX™, and SYBR™ dyes on Applied Biosystems™ QuantStudio™ 3 and 5 96-well 0.1mL RT-PCR systems.

It enables simultaneous multiple dye calibrations in one plate, significantly reducing effort and time required for dye calibration. Please refer to the instrument user guide for full instructions on usage.

Specifications

Format 96-well, 0.1 mL
Content And Storage Store at –20°C ± 5°C.

Calibrated Dye VIC
For Use With (Equipment) QuantStudio™ 3, QuantStudio™ 5, QuantStudio™ 6 Pro, QuantStudio™ 7 Pro, QuantStudio™ 6 Flex (Software version 1.6 or later), QuantStudio™ 7 Flex (Software version 1.6 or later)
Product Line FAM, QuantStudio, ROX, SYBR, VIC
Product Type Calibration Plate
For Use With (Application) Spectral Calibration
Shipping Condition Dry Ice
Quantity 1 plate
Plate Compatibility 96-well, 0.1 mL
Calibration Type Spectral Calibration
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.