Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Applied Biosystems™ QuantStudio™ 12K Flex OpenArray™ Plate Press 2.0

Catalog No. A24945
Encompass
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
1 each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24945 1 each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. A24945 missing translation for 'mfr' Applied Biosystems™ missing translation for 'supplierNo' A24945

Used to align and seal the QuantStudio OpenArray plate case and lid after sample loading.

Once sealed, the case is filled with immersion fluid and run on the QuantStudio™ 12K Flex Real-Time PCR System. A built-in timer with status indicator and a robust design provides for uniform and reproducible use.

Specifications

For Use With (Equipment) QuantStudio™ Openarray Plate Case
Product Line GTK- TAQMAN OPENARRAY GT ACC'S, OpenArray
Product Type Plate Press
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Room Temperature
Quantity 1 each
Plate Compatibility  OpenArray™ Plates,  TaqMan™ OpenArray™ Genotyping Plates
Color Gray

For Research Use Only. Not for use in diagnostic procedures.