missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ QuantStudio™ 12K Flex Accufill System

Catalog No. 4471021
Encompass
Change view
Click to view available options
Quantity:
1 system
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
4471021 1 system
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 4471021 Supplier Applied Biosystems™ Supplier No. 4471021

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Includes

Sample-filling System

orderInfo,Quantity,ItemDescription,ThermalblockFormats,HeightMetric,WidthMetric,DepthMetric,DynamicRange,Multiplex,Voltage,Detection,VolumeMetricThermalBlockSample,Sensitivity,WeightEnglish,WeightMetric,ForUseWithApplication

The QuantStudio™ 12K Flex Real-Time PCR System includes easy-to-change blocks that can accommodate OpenArray™ plates, TaqMan™ array cards, 384-well, 96-well, and FAST 96-well plates allowing you to select the format that is right for your project whether it’s now or in the future, for maximum flexibility and consistency in your research. The QuantStudio™ 12K Flex System’s intuitive software, easy touch screen setup, automation integration, effortless block exchange, and easy-to- use OpenArray™ components are designed to let you get started right away and reduce hands on time. Plan for the future and save time, money, and space in your lab with the only all-in-one qPCR instrument enabling a broad range of applications with maximum throughput.

Flexibility & Scalability:
  • Includes five interchangeable blocks: The QuantStudio™ 12K Flex blocks accommodate 96-well, FAST 96-well, 384-well, TaqMan™ array cards, and OpenArray™ plates, for effortless scaling from 1 to 12,000 data points in a single run. Increase throughput as you dive deeper into your research, using just one instrument. As you begin your project, start with 96-well format experiments and then scale up to a higher throughput to get the statistically significant answers you need. Additionally, you can easily validate and screen your targets of interest with a selected panel of genes using novel formats such as the TaqMan™ array cards and OpenArray™ plates.
  • Compatible with a full range of TaqMan™ assays: The QuantStudio™ 12K Flex System is optimized to enable clear, clean data for all TaqMan™ assays that might be used across a broad range of applications, including gene expression, microRNA and small noncoding RNAs, SNP genotyping, protein analysis, protein thermal shift, high resolution melt, copy number variation, and pathogen detection.
  • Delivers peace of mind: Leave early on a Friday and monitor runs from home, knowing that the system can safely store over 100 runs.

Maximum Throughput:
  • Enhanced OptiFlex™ System: Building upon Applied Biosystems’ proprietary technology, the Enhanced OptiFlex™ System provides enhanced fluorescence detection for the accurate and sensitive data analysis necessary to collect 12,000 data points in a single run.
  • Unprecedented coverage and sample throughput: The OpenArray™ format accelerates genomic validation and screening, driving over 150,000 data points per work day, enabling most projects to be completed in days instead of weeks.
  • Simple OpenArray™ workflow: QuantStudio™ Open Array™ plates arrive in a case to facilitate ease of handling and ensure optimal data quality upon loading of samples using the AccuFill™ System. Easily load four arrays into the QuantStudio™ 12K Flex instrument to achieve over 12,000 data points. Integrated sample tracking software lets you easily track your samples throughout the course of your project.
  • Integrated analysis and quality systems: New intuitive touch-screen interface and integrated calendar feature allows for scheduling time of use on the instrument. Additional analysis software such as ExpressionSuite v1.0 and Genotyper v1.1 is available to help you analyze hundreds of targets and samples and achieve the statistical power you need.

QuantStudio™ 12K Flex Software:
  • Intuitive interface and innovative design for novice and experienced users
  • Convenient walk-away operation
  • Improved data analysis across block types
  • Fully compatible with high-throughput environments

Expandable Software Architecture:
Optional add-ons are available to upgrade the existing software:
  • 21 CFR Part 11 compliance: This optional SAE module assists with Security, Auditing, and Electronic signature of records for full data traceability.
  • High Resolution Melt (HRM): Using the built-in MeltDoctor™ protocols and calibrations, you can spend less time optimizing the run and more time customizing your results. Define the number of variants you want to see by quickly changing analysis settings.
  • ExpressionSuite Software v1.0: our latest stand-alone gene expression software with enhanced analysis tools that can analyze over 200 384-well plates or 10 OpenArray™ plates.

For Research Use Only. Not for use in diagnostics procedures.

Order Info

Room Temperature

Specifications

Format OpenArray™ Plate
Content And Storage This Instrument comes with the Accufill System, 5 Interchangeable Blocks
High-throughput Compatibility High-throughput Compatible
For Use With (Equipment) QuantStudio™
Product Line QuantStudio
Product Type Accufill System
For Use With (Application) Real Time PCR (qPCR)
Shipping Condition Room Temperature
Volume (Metric) Thermal Block Sample 33 nl Reactions (OpenArray™ Plates)
Quantity 1 system
What type of Real-Time PCR (Sequence Detection System) information is available from Applied Biosystems?

Tutorials demonstrating introductions to Real-Time PCR, guidelines for designing real-time assays using Primer Express software, and various instrument set-ups can be found under Life Technologies University on our website. User bulletins, manuals and Product inserts, protocols can be found as downloadable pdf files on our Technical Resources page.