Learn More
United Scientific™ Quantitative Electroscope
The quantitative electroscope is ideal for measuring the strength of an electrostatic charge.
Supplier:  United Scientific™ ELQUANT
Catalog No. S000856
Only null left
Explore available promotions
Alert:
Prop 65
Description
Description
A well-balanced vane rotates in response to an electrical charge. The angle measurements printed on the back show different positions in increments of 10° up to 90°. The stronger the charge the greater the deflection. The metal case has a diameter of 5 in. and is 3 in. deep to help reduce the influence of air currents. The top plate is 3 in. in diameter.
  • Wide walls to reduce the effects of air currents
  • Well-balanced and sensitive vane
  • Easy to read protractor
  • Includes activity guide
Specifications
Specifications
Electroscope
Product Suggestions
Product Suggestions