missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Quantikine Immunoassay Control Set 869 for Glucagon

Catalog No. QC100
Change view
Click to view available options
Quantity:
1 Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
QC100 1 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. QC100 Supplier R&D Systems™ Supplier No. QC100

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

For use with catalog number DGCG0

Quantikine Immunoassay Control Set 869 for Glucagon is a research-grade control for use in ELISA applications. The control is provided as a lyophilized white powder that requires reconstitution with deionized or distilled water.

  • Applications: Control, ELISA
  • Species Reactivity: Multi-Species
  • Related Gene: GCG
  • Storage: Store unopened product at 2°C to 8°C; stable for at least 6 months from date of receipt.

Specifications

Content And Storage Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Detectable Analytes Glucagon
Type Immunoassay Control
Intended Use Control
Product Line R&D Systems
Quantity 1 Each
Source Multi-species
Form Lyophilized