Learn More
Bioassay Systems QuantiChrom™ Lipase Assay Kit. For quantitative determination of lipase activity.
Supplier:  Bioassay Systems DLPS100
Encompass
For quantitative determination of lipase enzyme activity. Key Features: Sensitive and accurate. Linear detection range 40 to 1600 U/L lipase activity in 96-well plate assay. Convenient and high throughput. The procedure involves adding a single working reagent, and reading the optical density at 10 min and 20 min at room temperature or 37°C. Can be automated to process thousands of samples per day. Method: OD412nm. Samples: Serum, plasma, saliva, urine etc. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 40 U/L.
Catalog No. 50489228
Only null left