Learn More
Waters Limited QuanRecovery with MaxPeak HPS Vial Package (186009186) with pre-slit PTFE silicone cap and septum, 100/pk
Catalog No. NC1675367
Encompass
QuanRecovery Vials with MaxPeak HPS reduce peptide and protein losses due to ionic interactions and non specific binding. A reduction in these losses lead to higher recovery, sensitivity and repeatability in analytical results. These 300L vials are LC-MS autosampler ready, with low residual volumes to fully utilize low sample volumes. MaxPeak High Performance Surfaces are new and innovative technologies designed to increase analyte recovery, sensitivity and reproducibility by minimizing analyte/surface interactions that can lead to sample losses.
Catalog No. NC1675367 Supplier Waters Limited Supplier No. 176004434
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
QuanRecovery Vials with MaxPeak HPS reduce peptide and protein losses due to ionic interactions and non specific binding. A reduction in these losses lead to higher recovery, sensitivity and repeatability in analytical results. These 300L vials are LC-MS autosampler ready, with low residual volumes to fully utilize low sample volumes. MaxPeak High Performance Surfaces are new and innovative technologies designed to increase analyte recovery, sensitivity and reproducibility by minimizing analyte/surface interactions that can lead to sample losses.