Learn More
BINDER™ Qualification Documents IQ/OQ /PQ
Catalog No. 08111925
Change view
Click to view available options
For Use With (Equipment):
BINDER Chambers (Series BD, BF, ED, FD, FED, FDL, FP, M, C, CB, KB, KT, MK, MKT, VD, VDL, UF V)
BINDER Chambers (Series KBF P, KBF LQC, KBW, KBWF)
BINDER Chambers (Series KBF, KBF-S, KMF, MKF, MKFT)
Product Type:
IQ/OQ Documentation (Hardcopy)
IQ/OQ Documentation (PDF)
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type
08111925 BINDER Chambers (Series BD, BF, ED, FD, FED, FDL, FP, M, C, CB, KB, KT, MK, MKT, VD, VDL, UF V) IQ/OQ Documentation (Hardcopy)
08111919 BINDER Chambers (Series BD, BF, ED, FD, FED, FDL, FP, M, C, CB, KB, KT, MK, MKT, VD, VDL, UF V) IQ/OQ Documentation (PDF)
08111920 BINDER Chambers (Series KBF, KBF-S, KMF, MKF, MKFT) IQ/OQ Documentation (PDF)
08111921 BINDER Chambers (Series KBF P, KBF LQC, KBW, KBWF) IQ/OQ Documentation (PDF)
08111926 BINDER Chambers (Series KBF, KBF-S, KMF, MKF, MKFT) IQ/OQ Documentation (Hardcopy)
08111927 BINDER Chambers (Series KBF P, KBF LQC, KBW, KBWF) IQ/OQ Documentation (Hardcopy)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 08111925 Supplier BINDER™ Supplier No. 70070005
Only null left
Qualification documents - IQ/OQ/PQ documents – supporting documents for validation performed by customers, according to customer requirements, PQ section added to qualification folder IQ/OQ; parameters: temperature, CO2, O2 – or pressure, depending on unit.
  • Parameters depending on unit
  • Available as hardcopy inside folder or PDF document
Specifications
For Use With (Equipment) BINDER Chambers (Series BD, BF, ED, FD, FED, FDL, FP, M, C, CB, KB, KT, MK, MKT, VD, VDL, UF V)
Description Qualification documents - Hard copy inside folder, IQ/OQ/PQ documents - supporting documents for validation performed by customers, according to customer requirements, PQ section added to qualification folder IQ/OQ; parameters: temperature, CO2, O2 - or pressure, depending on unit
Duration On request
Product Type IQ/OQ Documentation (Hardcopy)
Service Offering IQ/OQ/PQ with Completion of Documents; Parameters: Temperature, CO2, O2, or Pressure, Depending on Unit