Learn More
Invitrogen™ Qualification Add-on For CL1000 Imaging System
Catalog No. ZGQUSCIBRCL
Encompass
Catalog No. ZGQUSCIBRCL Supplier Invitrogen™ Supplier No. ZGQUSCIBRIGHTCL
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Designed to help balance business and laboratory requirements and help comply with global standards, country-specific regulations, and other laboratory site requirements.
  • Verify instrument performance and minimize downtime with IQ/OQ services (Installation Qualification and Operation Qualification)
  • Complete with audit ready documentation, factory-trained and experienced engineers will conduct IQ/OQ services
  • Qualification services help ensure instruments are installed and operating according to manufacturer's specifications
  • Helps move into production faster
  • IQ/OQ recommended: At installation , after a move
  • OQ recommended: After planned maintenance, After major repair, After additions, changes, or upgrades to the system, For a scheduled OQ site requirement
Specifications
For Use With (Equipment) iBright FL1000 Imaging System
Product Type Qualification Add-on
For Use With (Application) Warranty for Western Blotting instrumentation
Includes Audit-ready documentation, Order and system verification, Documentation verification, Installation verification, Software verification, Hardware operation verification, Component verification, Service coverage verification, Customer training verification