missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QSY™ 7 Carboxylic Acid, Succinimidyl Ester

Catalog No. q10193
Encompass
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q10193 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. Q10193 Supplier Invitrogen™ Supplier No. Q10193
Only null left

QSY™ 7 carboxylic acid, succinimidyl ester

QSY-7 succinimidyl ester is a nonfluorescent acceptor dye for preparation of peptide and oligonucleotide FRET probes via aliphatic amine modification.

Specifications

Quantity 5 mg
Product Type Succinimidyl Ester
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, Succinimidyl Ester
Label or Dye QSY™ 7
Label Type Quencher Dyes
Product Line QSY

For Research Use Only. Not for use in diagnostic procedures.