missing translation for 'onlineSavingsMsg'
Learn More

QRP™ Qualatrile™ 9C Anti-Static Latex Finger Cots

Catalog No. p-3202041
Change view
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19076190A Small
19076190C Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19076190A Supplier QRP™ Supplier No. GP9CS
Only null left

Packaged for ISO 5 (Class 100) applications, ideal for use with Class II static sensitive devices.

  • For use with Class II static sensitive devices with thresholds above 1000 V
  • Packaged for ISO 5 (Class 100) clean room applications
  • Average surface resistance of 109 ohms/sq.
  • Stand-up pouch allows for easy product identification

Specifications

Chlorinated Yes
Cleanroom Class Class 100 (ISO 5)
Accelerator Free Yes
Applications Industrial
Color White
Disposable Single-use
Exterior Finish No Texture
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type ESD Finger Cot
Size Small
Sterility Sterile
Thickness (English) 3 mil
Thickness (Metric) 0.08 mm
For Use With (Application) Avionics, Consumer Electronics, IC Manufacturing, and Assembly/Repair Operations
For Use With (Equipment) Class II Static-Sensitive Devices
Packaging Type Stand-up Pouch
Length (English) 2.8 in.
Length (Metric) 7 cm
Show More Show Less