Learn More
Alkali Scientific qPCR/PCR Tube Rack w Lid, (BLUE) Fits 96 x 0.2mL Tubes, Strips or Plates, 5/pk
Catalog No. 502389005
Encompass
Organize and protect your 0.2mL tubes with this durable qPCR/PCR Tube Rack. This blue rack comes with a secure lid with alphanumeric indexing to ensure sample integrity and has 96 wells. Each pack includes 5 racks, making it perfect for high-throughput labs. Stay efficient and color-coordinated with this essential laboratory accessory.
Catalog No. 502389005 Supplier Alkali Scientific Supplier No. R9602B
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Organize and protect your 0.2mL tubes with this durable qPCR/PCR Tube Rack. This blue rack comes with a secure lid with alphanumeric indexing to ensure sample integrity and has 96 wells. Each pack includes 5 racks, making it perfect for high-throughput labs. Stay efficient and color-coordinated with this essential laboratory accessory.