missing translation for 'onlineSavingsMsg'
Learn More

Qorpak™ White Polypropylene Hole Caps with Bonded PTFE/Silicone Septa

Catalog No. p-4603627
Change view
Click to view available options
Neck Finish:
28-400
No. per Case:
1000 Cs.
Packaging:
Bulk
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Neck Finish No. per Case Packaging
02991476 28-400 1000 Cs. Bulk
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02991476 Supplier Qorpak™ Supplier No. CAP00343

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

No slipping away of septa from cap while injecting

  • Good impact strength with pliability
  • Septa features high resealing and chemical resistant properties

Specifications

Color White
Material Polypropylene
No. per Case 1000 Cs.
Packaging Bulk
Sterility Non-sterile
Liner Material PTFE/Silicone
Neck Finish 28-400
Disposable Reusable
Product Type Threaded Screw Cap
For Use With (Equipment) Wide mouth bottle
Autoclavable Not Autoclavable
Show More Show Less