missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Qorpak™ CurTec™ Wide Neck Drums
Provides high level of product protection and added value through quality, functionality, ease of use, and design. Qorpak™ CurTec™ Wide Neck Drums are used for shipping and stocking valuable fine chemicals.
$694.94 - $1,384.12
Specifications
| Product Type | Drum/Pail with Screw-on Lid |
|---|---|
| Certifications/Compliance | HACCP, ISO |
| Closure Type | Cap |
| Color | White |
| For Use With (Application) | Ideal for pharmaceuticals, fine chemicals, food ingredients and hazardous waste |
| Catalog Number | Mfr. No. | Diameter (English) Outer | Height (English) | Weight (English) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (English) Outer | Height (English) | Weight (English) | Price | Quantity & Availability | |||
|
02994113
|
Qorpak™
248723 |
6.5 in. | 7.7 in. | 21 lb. |
Case of 10 Each for $694.94
|
|
|||
|
02994114
|
Qorpak™
248725 |
7.7 in. | 10.3 in. | 30 lb. |
Case of 15 Each for $1,384.12
|
|
|||
|
02994117
|
Qorpak™
248733 |
10.7 in. | 16.4 in. | 32 lb. |
Case of 4 Each for $876.56
|
|
|||
Description
- Features red polypropylene unlined lid
- Wide neck for easy filling of contents
Certifications
HACCP, ISO
Specifications
| Drum/Pail with Screw-on Lid | |
| Cap | |
| Ideal for pharmaceuticals, fine chemicals, food ingredients and hazardous waste |
| HACCP, ISO | |
| White | |
| HDPE |