missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals QNZ, 98%

$128.85

Chemical Identifiers

CAS 545380-34-5
Molecular Formula C22H20N4O
Synonym 6-Amino-4-[2-(4-phenoxyphenyl)ethylamino]quinazoline;
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AAJ67487MCR
View Documents Promotion Details
Thermo Scientific Chemicals
J67487MCR
1 mg
Each for $128.85
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

545380-34-5
C22H20N4O
6-Amino-4-[2-(4-phenoxyphenyl)ethylamino]quinazoline;

Specifications

White
545380-34-5
356.43
QNZ, 98%
IBAKVEUZKHOWNG-UHFFFAOYSA-N
N4-[2-(4-phenoxyphenyl)ethyl]quinazoline-4,6-diamine
98%
-30°C to -10°C
C22H20N4O
6-Amino-4-[2-(4-phenoxyphenyl)ethylamino]quinazoline;
Solid
NC1=CC2=C(NCCC3=CC=C(OC4=CC=CC=C4)C=C3)N=CN=C2C=C1
356.43
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

RUO – Research Use Only