missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Axygen™ Qiagen™ Robotic Tips
Description
Available in volumes of 250, 300, 1000 and 1100 μL.
- Clear and conductive black tips available
- Choice of Presterilized, Nonsterilized, Filtered, Non-Filtered and Conductive tips
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Filtered | Filtered |
| Sterility | Sterile |
| Color | Clear |
| Certifications/Compliance | Non-ANSI |
| Clean Claims | DNase, Rnase Free |
| For Use With (Application) | Non-pyrogenic, used for stringent specifications under strict process controls |
| Format | Rack |
| Tip Style | Automation, Round |
| For Use With (Equipment) | Qiagen BioRobot |
| Show More |