missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ Qiagen™ Robotic Tips

Catalog No. 14222388
Change view
Click to view available options
Volume (Metric):
1000 μL
Sterility:
Sterile
Color:
Clear
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Sterility Color
14222388 1000 μL Sterile Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14222388 Supplier Axygen™ Supplier No. 4345
Get Up to $2,500 in Products with Purchase
View all promotions

missing translation for 'validMainframeMsg'

Compatible with Qiagen liquid handling systems and Qiagen BioRobot Series

Available in volumes of 250, 300, 1000 and 1100 μL.

  • Clear and conductive black tips available
  • Choice of Presterilized, Nonsterilized, Filtered, Non-Filtered and Conductive tips

Specifications

Volume (Metric) 1000 μL
Filtered Filtered
Sterility Sterile
Color Clear
Certifications/Compliance Non-ANSI
Clean Claims DNase, Rnase Free
For Use With (Application) Non-pyrogenic, used for stringent specifications under strict process controls
Format Rack
Tip Style Automation, Round
For Use With (Equipment) Qiagen BioRobot
Material Polypropylene
Show More Show Less