Learn More
Tosoh Bioscience Llc QC-PAK GFC 300 7.8X150 DFS Item
Supplier:  Tosoh Bioscience Gmbh TB16049
Column, Gel Filtration Chromatography; Tosoh TSKgel QC-PAK GFC 300; Media: Silica; Particle: 5 micrometer; Pore Size: 250 angstrom; 7.8mm I.D. x 15cm Length; Stainless Steel
Catalog No. 50851380
Only null left
Explore available promotions