Learn More
QBC Diagnostics Autoread™ Plus Dry Centrifugal Hematology System
Autoread™ Plus Dry Centrifugal Hematology System for cost-effective and maintenance free hematology.
$2,843.07
Specifications
| For Use With (Application) | Dry hematology technology using tubes that are internally coated with necessary reagents |
|---|---|
| Certifications/Compliance | CLIA EQC Option 2 |
| Electrical Requirements | 120/230V, 50/60Hz |
| Frequency | 50/60 Hz |
| Voltage | 120/230 V |
Description
- Employs dry hematology technology using tubes that are internally coated with necessary reagents—no need for costly liquid reagents.
- AccuTubes can be used for both venous and capillary blood samples.
- Yields quantitative values from venous or capillary blood sample for nine parameters:
- Hematocrit (HCT)
- Hemoglobin (HgB)
- Mean Corpuscular Hemoglobin Concentration (MCHC)
- White Blood Cell Count (WBC)
- Granulocyte Count/Percentage
- Lymphocyte and Monocyte Count/Percentage
- No routine maintenance or calibration necessary—a simple calibration check is run daily on a calibration rod to verify performance.
- Liquid controls available separately for additional performance monitoring.
- Optical Signature Analysis automatically scans collection tube eight times, eliminating subjective interpretation.
- Printouts can include age-and sex-specific normal ranges and error messages to alert user to improper tube preparation, unusual sample results, or other irregular conditions.
- Analyzer software contains hematology diagnostic reminders (HDR)
- Users may select from several printed formats for test results:
- Test values only
- Test values plus abbreviated HDR comments
- Test values plus expanded HDR comments
Compliance
Complies with CLIA EQC Option 2.
Specifications
| Dry hematology technology using tubes that are internally coated with necessary reagents | |
| 120/230V, 50/60Hz | |
| 120/230 V |
| CLIA EQC Option 2 | |
| 50/60 Hz |