Learn More
MilliporeSigma™ Q-POD™ Remote Dispenser for Ultrapure Water
Catalog No. ZIQP0D000
Change view
Click to view available options
Type:
Q-POD™ Remote Dispenser for Ultrapure Water
Catalog No. Type
ZIQP0D000 Q-POD™ Remote Dispenser for Ultrapure Water
1 options
Catalog No. ZIQP0D000 Supplier MilliporeSigma™ Supplier No. ZIQP0D000
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Intelligent ultrapure water dispenser. Up to 4 Q-POD™s can be connected to a Milli-Q™ IQ 7000 water purification system.
Our Lab Water purification system portfolio offers a broad range of pure and ultrapure water purification systems and services designed for scientists working in pharmaceutical, clinical, academic, industrial, research, and government laboratories — in both validated and non-validated environments. 
Specifications
Type Q-POD™ Remote Dispenser for Ultrapure Water
For Use With (Equipment) Milli-Q IQ 7000 Ultrapure Water System
Height (English) 27.6 in.
Height (Metric) 70.2 cm