missing translation for 'onlineSavingsMsg'
Learn More

Pyruvonitrile, tech. 90%

Catalog No. p-7057206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6663603 1 g
AAH6663606 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6663603 Supplier Thermo Scientific Chemicals Supplier No. H6663603
Only null left

CAS: 631-57-2 | C3H3NO | 69.063 g/mol

Pyruvonitrile was used in the synthesis of cuprate-carbonyl π-complexes by reaction with Me2CuLi in THF.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyruvonitrile was used in the synthesis of cuprate-carbonyl π-complexes by reaction with Me2CuLi in THF.

Solubility
Soluble in ether and acetonitrile.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.

Chemical Identifiers

CAS 631-57-2
Molecular Formula C3H3NO
Molecular Weight (g/mol) 69.063
MDL Number MFCD00001838
InChI Key QLDHWVVRQCGZLE-UHFFFAOYSA-N
Synonym pyruvonitrile, propanenitrile, 2-oxo, oxopropionitrile, 2-oxopropiononitrile, 2-oxopropionitrile, oxypropionitrile, pyruvic acid nitrile, unii-ulo441649v, accn, .alpha.-oxopropionitrile
PubChem CID 69430
IUPAC Name acetyl cyanide
SMILES CC(=O)C#N

Specifications

Color Colorless to Brown
Density 0.974 g/mL
Boiling Point 92°C to 93°C
Flash Point 14°C (57°F)
Refractive Index 1.3755
Linear Formula CH3COCN
Quantity 1 g
UN Number UN3273
Beilstein 1737633
Solubility Information Soluble in ether and acetonitrile.
Formula Weight 69.06
Percent Purity 90%
Grade Technical
Physical Form Liquid
Chemical Name or Material Pyruvonitrile
Show More Show Less
Hazard Category H225-H301-H315-H319-H330-H335
Hazard Statement GHS H Statement
H225-H300-H330-H315-H335
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal if inhaled.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P284-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packaging Group: II; Proper Shipping Name: NITRILES, FLAMMABLE, TOXIC, N.O.S.
EINECSNumber 211-159-2
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only