missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic Aldehyde Dimethyl Acetal 97.0+%, TCI America™

Catalog No. P126425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P126425ML 25 mL
P1264100ML 100 mL
P1264500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. P126425ML Supplier TCI America Supplier No. P126425ML

Chemical Identifiers

CAS 6342-56-9
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.132
MDL Number MFCD00008758
InChI Key ULVSHNOGEVXRDR-UHFFFAOYSA-N
Synonym 1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal
PubChem CID 80650
IUPAC Name 1,1-dimethoxypropan-2-one
SMILES CC(=O)C(OC)OC

Specifications

Boiling Point 141°C
Quantity 25 mL
UN Number 1993
Formula Weight 118.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Pyruvic Aldehyde Dimethyl Acetal