missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic aldehyde dimethyl acetal, 98%

CAS: 6342-56-9 | C5H10O3 | 118.13 g/mol

$86.40 - $86.40

Chemical Identifiers

CAS 6342-56-9
Molecular Formula C5H10O3
Molecular Weight (g/mol) 118.13
MDL Number MFCD00008758
InChI Key ULVSHNOGEVXRDR-UHFFFAOYSA-N
Synonym 1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal
PubChem CID 80650
IUPAC Name 1,1-dimethoxypropan-2-one
SMILES CC(=O)C(OC)OC
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC173801000
SDS View Documents
Thermo Scientific Chemicals
173801000
100 g
Each for $86.40
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

6342-56-9
118.13
ULVSHNOGEVXRDR-UHFFFAOYSA-N
80650
CC(=O)C(OC)OC
C5H10O3
MFCD00008758
1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal
1,1-dimethoxypropan-2-one

Specifications

6342-56-9
Colorless to Yellow
143.0°C to 147.0°C
Authentic
Glass bottle
1.3970 to 1.3990
MFCD00008758
01, IV, 3631
1,1-dimethoxyacetone, pyruvic aldehyde dimethyl acetal, 2-propanone, 1,1-dimethoxy, methylglyoxal dimethyl acetal, 1,1-dimethoxy-2-propanone, dimethoxymethyl methyl ketone, pyruvaldehyde dimethyl acetal, 1,1-dimethoxy acetone, dimethoxyacetone, pyruvaldehyde, 1-dimethyl acetal
ULVSHNOGEVXRDR-UHFFFAOYSA-N
1,1-dimethoxypropan-2-one
80650
118.13
Liquid
-57.0°C
1.0000g/mL
37°C
97.5% min. (GC)
C5H10O3
CH3COCH(OCH3)2
100 g
1
Solubility in water: 350g/L (20°C). Other solubilities: miscible with methanol, acetone and thf
CC(=O)C(OC)OC
118.13
0.85 mPa.s (25°C)
98%
Pyruvic aldehyde dimethyl acetal, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 228-735-4

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only