missing translation for 'onlineSavingsMsg'
Learn More

Pyruvic acid, sodium salt, 99+%

Catalog No. p-3737306
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC132150250 25 g Plastic bottle
AC132151000 100 g Plastic bottle
AC132155000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC132150250 Supplier Thermo Scientific Chemicals Supplier No. 132150250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 113-24-6 | C3H3NaO3 | 110.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 113-24-6
Molecular Formula C3H3NaO3
Molecular Weight (g/mol) 110.04
MDL Number MFCD00002586
InChI Key DAEPDZWVDSPTHF-UHFFFAOYSA-M
Synonym sodium pyruvate, pyruvic acid sodium salt, sodium 2-oxopropanoate, pyruvic acid, sodium salt, propanoic acid, 2-oxo-, sodium salt, sodium alpha-ketopropionate, pyruvate sodium, unii-pod38aif08, 2-oxopropanoic acid sodium salt, pod38aif08
PubChem CID 23662274
ChEBI CHEBI:50144
IUPAC Name sodium;2-oxopropanoate
SMILES CC(=O)C(=O)[O-].[Na+]

Specifications

Melting Point >300.0°C
Color Colorless to Yellow
pH 5.5 to 6.5 (10% soln. at 20°C)
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula CH3COCO2Na
Quantity 25 g
Beilstein 03, 608
Solubility Information (10% in water) Clear colorless to light yellow
Formula Weight 110.04
Percent Purity 99+%
Physical Form Solution
Chemical Name or Material Pyruvic acid, sodium salt
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 204-024-4

RUO – Research Use Only