missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrrolidine, TCI America™
$138.61 - $138.61
Chemical Identifiers
| CAS | 123-75-1 |
|---|---|
| Molecular Formula | C4H10ClN |
| Molecular Weight (g/mol) | 107.58 |
| MDL Number | MFCD00005249 |
| InChI Key | FCLZCOCSZQNREK-UHFFFAOYSA-N |
| Synonym | tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine |
| PubChem CID | 31268 |
| ChEBI | CHEBI:33135 |
| IUPAC Name | hydrogen pyrrolidine chloride |
| SMILES | [H+].[Cl-].C1CCNC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
P0576500ML
|
TCI America
P0576500ML |
500 mL |
N/A
|
|
|||||
Chemical Identifiers
| 123-75-1 | |
| 107.58 | |
| FCLZCOCSZQNREK-UHFFFAOYSA-N | |
| 31268 | |
| hydrogen pyrrolidine chloride |
| C4H10ClN | |
| MFCD00005249 | |
| tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine | |
| CHEBI:33135 | |
| [H+].[Cl-].C1CCNC1 |
Specifications
| 123-75-1 | |
| Colorless | |
| C4H10ClN | |
| 500 mL | |
| tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine | |
| [H+].[Cl-].C1CCNC1 | |
| 107.58 | |
| CHEBI:33135 | |
| ≥98.0% (GC) | |
| Pyrrolidine |
| -63°C | |
| 88°C | |
| MFCD00005249 | |
| 1922 | |
| FCLZCOCSZQNREK-UHFFFAOYSA-N | |
| hydrogen pyrrolidine chloride | |
| 31268 | |
| 71.12 | |
| Liquid |
Safety and Handling
EINECSNumber : (5)-0103
RTECSNumber : UX9650000
TSCA : Yes