missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine, TCI America™

Catalog No. P0576500ML
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P0576500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P0576500ML Supplier TCI America Supplier No. P0576500ML

Chemical Identifiers

CAS 123-75-1
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.12
MDL Number MFCD00005249
InChI Key RWRDLPDLKQPQOW-UHFFFAOYSA-N
Synonym tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
PubChem CID 31268
ChEBI CHEBI:33135
IUPAC Name pyrrolidine
SMILES C1CCNC1

Specifications

Melting Point -63°C
Color Colorless
Boiling Point 88°C
Quantity 500 mL
UN Number 1922
Formula Weight 71.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Pyrrolidine
EINECSNumber (5)-0103
RTECSNumber UX9650000
TSCA Yes