missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine Hydrobromide 98.0+%, TCI America™

Catalog No. P24845G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P24845G 5 g
P24841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P24845G Supplier TCI America Supplier No. P24845G

Chemical Identifiers

CAS 55810-80-5
Molecular Formula C4H10BrN
Molecular Weight (g/mol) 152.035
InChI Key VFDOIPKMSSDMCV-UHFFFAOYSA-N
Synonym Pyrrolidinium Bromide
PubChem CID 18621471
IUPAC Name pyrrolidine;hydrobromide
SMILES C1CCNC1.Br

Specifications

Quantity 5 g
Formula Weight 152.04
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrrolidine Hydrobromide