missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrrolidine Hydriodide 98.0+%, TCI America™
Specifications
Specifications
| Quantity | 1 g |
| Formula Weight | 199.04 |
| Percent Purity | ≥98.0% (N,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Pyrrolidine Hydriodide |
Safety and Handling
| TSCA | No |