missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™

CAS: 123-75-1 | C4H10ClN | 107.58 g/mol

$127.77 - $127.77

Chemical Identifiers

CAS 123-75-1
Molecular Formula C4H10ClN
Molecular Weight (g/mol) 107.58
MDL Number MFCD00005249
InChI Key FCLZCOCSZQNREK-UHFFFAOYSA-N
Synonym tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
PubChem CID 31268
ChEBI CHEBI:33135
IUPAC Name hydrogen pyrrolidine chloride
SMILES [H+].[Cl-].C1CCNC1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC446141000
SDS View Documents
Thermo Scientific Chemicals
446141000
100 mL AcroSeal™ Glass bottle
Each for $127.77
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

123-75-1
107.58
FCLZCOCSZQNREK-UHFFFAOYSA-N
31268
hydrogen pyrrolidine chloride
C4H10ClN
MFCD00005249
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
CHEBI:33135
[H+].[Cl-].C1CCNC1

Specifications

123-75-1
0.8660g/mL
3°C
AcroSeal™ Glass bottle
1.4421 to 1.4441
100 mL
15,8129
Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether and chloroform, soluble in most organic solvents
[H+].[Cl-].C1CCNC1
107.58
0.94 mPa.s (20°C)
71.11
Pyrrolidine
-63.0°C
86.0°C to 88.0°C
99.4% min. (GC)
C4H10ClN
MFCD00005249
0.866
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
FCLZCOCSZQNREK-UHFFFAOYSA-N
hydrogen pyrrolidine chloride
31268
CHEBI:33135
99.5+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 204-648-7

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only