missing translation for 'onlineSavingsMsg'
Learn More

Pyrrolidine, 99.5+%, purified by redistillation, AcroSeal™

Catalog No. AC446141000
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC446141000 100 mL AcroSeal™ Glass bottle
AC446148000 800 mL AcroSeal™ Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC446141000 Supplier Thermo Scientific Chemicals Supplier No. 446141000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 123-75-1 | C4H10ClN | 107.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 123-75-1
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.12
MDL Number MFCD00005249
InChI Key RWRDLPDLKQPQOW-UHFFFAOYSA-N
Synonym tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine
PubChem CID 31268
ChEBI CHEBI:33135
IUPAC Name pyrrolidine
SMILES C1CCNC1

Specifications

Melting Point -63.0°C
Density 0.8660g/mL
Boiling Point 86.0°C to 88.0°C
Flash Point 3°C
Assay Percent Range 99.4% min. (GC)
Packaging AcroSeal™ Glass bottle
Refractive Index 1.4421 to 1.4441
Quantity 100 mL
Merck Index 15,8129
Specific Gravity 0.866
Solubility Information Solubility in water: freely soluble. Other solubilities: soluble in alcohol, ether and chloroform, soluble in most organic solvents
Viscosity 0.94 mPa.s (20°C)
Formula Weight 71.11
Percent Purity 99.5+%
Chemical Name or Material Pyrrolidine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful if inhaled.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 204-648-7

RUO – Research Use Only