Learn More
Pyrrolidine, 99%
CAS: 123-75-1 | C4H10ClN | 107.58 g/mol
$75.70 - $526.16
Chemical Identifiers
CAS | 123-75-1 |
---|---|
Molecular Formula | C4H10ClN |
Molecular Weight (g/mol) | 107.58 |
MDL Number | MFCD00005249 |
InChI Key | FCLZCOCSZQNREK-UHFFFAOYSA-N |
Synonym | tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimineShow More |
PubChem CID | 31268 |
ChEBI | CHEBI:33135 |
IUPAC Name | hydrogen pyrrolidine chloride |
SMILES | [H+].[Cl-].C1CCNC1 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA14852AE
|
Thermo Scientific Chemicals
A14852AE |
100 mL |
Each for $75.70
|
|
||||||
AAA14852AP
|
Thermo Scientific Chemicals
A14852AP |
500 mL |
Each for $159.64
|
|
||||||
AAA148520F
|
Thermo Scientific Chemicals
A148520F |
2500 mL |
Each for $526.16
|
|
||||||
Description
Pyrrolidine is used as building block in the synthesis of organic compounds. It is used to activate ketones and aldehydes toward nucleophilic addition. It is also utilized in promoting aldol condensation of ketones and aldehydes by forming enamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsPyrrolidine is used as building block in the synthesis of organic compounds. It is used to activate ketones and aldehydes toward nucleophilic addition. It is also utilized in promoting aldol condensation of ketones and aldehydes by forming enamine.
Solubility
Miscible with alcohol, ether, chloroform and water.
Notes
Store in cool place. Incompatible with acid chlorides, acid anhydrides, strong oxidizing agents, carbon dioxide and acids.
Chemical Identifiers
123-75-1 | |
107.58 | |
FCLZCOCSZQNREK-UHFFFAOYSA-N | |
31268 | |
hydrogen pyrrolidine chloride |
C4H10ClN | |
MFCD00005249 | |
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimineShow More | |
CHEBI:33135 | |
[H+].[Cl-].C1CCNC1 |
Specifications
123-75-1 | |
0.856 | |
2°C (35°F) | |
Like Rotten Eggs | |
1.4431 | |
100 mL | |
102395 | |
14,8015 | |
Miscible with alcohol,ether,chloroform and water. | |
[H+].[Cl-].C1CCNC1 | |
107.58 | |
CHEBI:33135 | |
99% | |
Pyrrolidine |
-63°C | |
86°C to 88°C | |
99% | |
C4H10ClN | |
MFCD00005249 | |
UN1922 | |
Air Sensitive | |
tetrahydropyrrole, tetrahydro pyrrole, azacyclopentane, azolidine, tetramethylenimine, butylenimine, perhydropyrrole, prolamine, 1-azacyclopentane, tetramethyleneimine | |
FCLZCOCSZQNREK-UHFFFAOYSA-N | |
hydrogen pyrrolidine chloride | |
31268 | |
71.12 | |
Liquid |
Safety and Handling
GHS H Statement
H225-H301-H314-H318-H332
Highly flammable liquid and vapour.
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if inhaled.
P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H225-H302+H332-H314
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PYRROLIDINE
EINECSNumber : 204-648-7
RTECSNumber : UX9650000
TSCA : Yes
storageNote1 : Air sensitive
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only