missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole 99.0+%, TCI America™

Supplier:  TCI America P0574500ML

 View more versions of this product

Catalog No. P0574500ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Pyrrole
-23°C
130°C
MFCD00005216
1992
KAESVJOAVNADME-UHFFFAOYSA-N
1H-pyrrole
8027
67.09
Liquid
109-97-7
Yellow
C4H5N
500 mL
pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c
N1C=CC=C1
67.09
CHEBI:19203
≥99.0% (GC)

Chemical Identifiers

109-97-7
67.09
KAESVJOAVNADME-UHFFFAOYSA-N
8027
1H-pyrrole
C4H5N
MFCD00005216
pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c
CHEBI:19203
N1C=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0100

RTECSNumber : UX9275000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions