missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole, 98+%

Catalog No. AAA1261618
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1261618 50 g
AAA1261630 250 g
AAA126160B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1261618 Supplier Thermo Scientific Chemicals Supplier No. A1261618

CAS: 109-97-7 | C4H5N | 67.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Pyrrole plays a major role in synthesis of drugs, spices, agrochemicals, dyes, photographic chemicals and perfumes. It plays an important role in the electropolymerisation of macroporous conducting polymer films. It acts as a catalyst for polymerization process; as a standard substance in chromatographic analysis; as corrosion inhibitors and preservatives and as solvents for resins and terpenes. It is utilized to study the hydrogen-bond mediated coupling of 1,2,3-triazole to pyrrole and in the preparation of 1-(4-Chloro-benzoyl)-pyrrole by reacting with 4-Chloro-benzoyl chloride. In Ciamician-Dennstedt rearrangement, It is used to prepare 3-chloropyridine by reacting with dichlorocarbene.

Solubility
Miscible with alcohol, ethyl ether, dilute acids and organic solvents. Immiscible with water.

Notes
Air, light and moisture sensitive. Store in cool place. Incompatible with strong acids, strong oxidizing agents, acid chlorides and acid anhydrides.

Chemical Identifiers

CAS 109-97-7
Molecular Formula C4H5N
Molecular Weight (g/mol) 67.09
MDL Number MFCD00005216
InChI Key KAESVJOAVNADME-UHFFFAOYSA-N
Synonym pyrrole, divinylenimine, azole, imidole, pyrrol, monopyrrole, divinyleneimine, 1-aza-2,4-cyclopentadiene, polypyrrole, unii-86s1zd6l2c
PubChem CID 8027
ChEBI CHEBI:19203
IUPAC Name 1H-pyrrole
SMILES N1C=CC=C1

Specifications

Density 0.967
Boiling Point 131°C
Flash Point 38°C (100°F)
Odor Mild
Refractive Index 1.509
Quantity 50 g
UN Number UN1992
Beilstein 1159
Sensitivity Air and light sensitive
Merck Index 14,8014
Solubility Information Miscible with alcohol,ethyl ether,dilute acids and organic solvents. Immiscible with water.
Formula Weight 67.09
Percent Purity ≥98%
Chemical Name or Material Pyrrole
Show More Show Less
Hazard Category H226-H301-H318-H332
Hazard Statement GHS H Statement
H301-H331-H318-H226
Toxic if swallowed.
Toxic if inhaled.
Causes serious eye damage.
Flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 203-724-7
RTECSNumber UX9275000
TSCA Yes
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only