missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-3-carboxylic acid hydrate, 95%

CAS: 336100-46-0 | C5H5NO2 | 111.10 g/mol

$187.80 - $187.80

Chemical Identifiers

CAS 336100-46-0
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.10
MDL Number MFCD06201862
InChI Key DOYOPBSXEIZLRE-UHFFFAOYSA-N
Synonym pyrrole-3-carboxylic acid hydrate, 1h-pyrrole-3-carboxylic acid hydrate, c5h5no2.h2o, 1h-pyrrole-3-carboxylicacid, hydrate 1:?
PubChem CID 45076181
IUPAC Name 1H-pyrrole-3-carboxylic acid;hydrate
SMILES OC(=O)C1=CNC=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC377655000
SDS View Documents
Thermo Scientific Chemicals
377655000
500 mg Glass bottle N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

336100-46-0
111.10
DOYOPBSXEIZLRE-UHFFFAOYSA-N
45076181
OC(=O)C1=CNC=C1
C5H5NO2
MFCD06201862
pyrrole-3-carboxylic acid hydrate, 1h-pyrrole-3-carboxylic acid hydrate, c5h5no2.h2o, 1h-pyrrole-3-carboxylicacid, hydrate 1:?
1H-pyrrole-3-carboxylic acid;hydrate

Specifications

336100-46-0
Beige to Brown
94% min. (HPLC)
C5H5NO2
500 mg
pyrrole-3-carboxylic acid hydrate, 1h-pyrrole-3-carboxylic acid hydrate, c5h5no2.h2o, 1h-pyrrole-3-carboxylicacid, hydrate 1:?
DOYOPBSXEIZLRE-UHFFFAOYSA-N
1H-pyrrole-3-carboxylic acid;hydrate
45076181
95%
Pyrrole-3-carboxylic acid hydrate
144.0°C to 147.0°C
Authentic
Glass bottle
MFCD06201862
22,II,17
Solubility in water: soluble. Other solubilities: soluble in benzene, diethyl ether, petrol. ether
OC(=O)C1=CNC=C1
111.10
111.1
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only