missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-3-carboxylic acid, 98+%

Catalog No. p-7036914
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH55383MD 250 mg
AAH5538303 1 g
AAH5538306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH55383MD Supplier Thermo Scientific Chemicals Supplier No. H55383MD

CAS: 931-03-3 | C5H5NO2 | 111.1 g/mol

Pyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.

Solubility
Soluble in water.

Notes
Incompatible with strong bases, strong oxidizing agents, strong reducing agents.

Chemical Identifiers

CAS 931-03-3
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00800594
InChI Key DOYOPBSXEIZLRE-UHFFFAOYSA-N
Synonym pyrrole-3-carboxylic acid, pyrrole-3-carboxylicacid, 3-carboxy-1h-pyrrole, 1h-pyrrole-3-carboxylicacid, pubchem8345, 3-carboxypyrrole, 3-pyrrolecarboxylic acid, acmc-1agqb, 3-pyrrole-carboxylic acid, pyrrole 3-carboxylic acid
PubChem CID 101030
ChEBI CHEBI:68076
IUPAC Name 1H-pyrrole-3-carboxylic acid
SMILES C1=CNC=C1C(=O)O

Specifications

Quantity 250 mg
Beilstein 108642
Solubility Information Soluble in water.
Formula Weight 111.1
Percent Purity ≥98%
Chemical Name or Material Pyrrole-3-carboxylic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only