missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-2-carboxylic Acid 98.0+%, TCI America™

Catalog No. p-7135044
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12701G 1 g
P12705G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12701G Supplier TCI America Supplier No. P12701G
Only null left

Chemical Identifiers

CAS 634-97-9
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00005219
InChI Key WRHZVMBBRYBTKZ-UHFFFAOYSA-N
Synonym pyrrole-2-carboxylic acid, minaline, minalin, 2-pyrrolecarboxylic acid, pyrrole-2-carboxylicacid, pyrrole-2-carboxylate, 2-carboxypyrrole, 1h-pyrrolecarboxylic acid, 9ci, pyc
PubChem CID 12473
ChEBI CHEBI:36751
IUPAC Name 1H-pyrrole-2-carboxylic acid
SMILES C1=CNC(=C1)C(=O)O

Specifications

Melting Point 208°C
Quantity 1 g
Formula Weight 111.10
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Pyrrole-2-carboxylic Acid