missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-2-carboxylic acid, 97%

Catalog No. AAL1375603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1375603 1 g
AAL1375606 5 g
AAL1375614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1375603 Supplier Thermo Scientific Chemicals Supplier No. L1375603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 634-97-9 | C5H5NO2 | 111.1 g/mol

Pyrrole-2-carboxylic acid has demonstrated antiparasitic activity against Trypanosomes by selective proline racemase inhibition and has potent antifungal activity against Phytophthora. It also has antifungal properties.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Pyrrole-2-carboxylic acid has demonstrated antiparasitic activity against Trypanosomes by selective proline racemase inhibition and has potent antifungal activity against Phytophthora. It also has antifungal properties.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Chemical Identifiers

CAS 634-97-9
Molecular Formula C5H5NO2
Molecular Weight (g/mol) 111.1
MDL Number MFCD00005219
InChI Key WRHZVMBBRYBTKZ-UHFFFAOYSA-N
Synonym pyrrole-2-carboxylic acid, minaline, minalin, 2-pyrrolecarboxylic acid, pyrrole-2-carboxylicacid, pyrrole-2-carboxylate, 2-carboxypyrrole, 1h-pyrrolecarboxylic acid, 9ci, pyc
PubChem CID 12473
ChEBI CHEBI:36751
IUPAC Name 1H-pyrrole-2-carboxylic acid
SMILES C1=CNC(=C1)C(=O)O

Specifications

Melting Point ∼205°C (sublimation)
Quantity 1 g
Beilstein 80825
Solubility Information Soluble in methanol.
Formula Weight 111.1
Percent Purity 99%
Chemical Name or Material Pyrrole-2-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-221-9
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only