missing translation for 'onlineSavingsMsg'
Learn More

Pyrrole-2-carboxaldehyde 98.0+%, TCI America™

Catalog No. p-7135028
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P12465G 5 g
P124625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P12465G Supplier TCI America Supplier No. P12465G
Only null left

Chemical Identifiers

CAS 1003-29-8
Molecular Formula C5H5NO
Molecular Weight (g/mol) 95.101
MDL Number MFCD00005217
InChI Key ZSKGQVFRTSEPJT-UHFFFAOYSA-N
Synonym pyrrole-2-carboxaldehyde, 2-formylpyrrole, 1h-pyrrole-2-carboxaldehyde, pyrrole-2-aldehyde, 2-pyrrolecarboxaldehyde, pyrrole-2-carbaldehyde, 2-pyrrolecarbaldehyde, 2-pyrrolylcarboxaldehyde, 2-pyrrolaldehyde, alpha-pyrrolaldehyde
PubChem CID 13854
ChEBI CHEBI:59978
IUPAC Name 1H-pyrrole-2-carbaldehyde
SMILES C1=CNC(=C1)C=O

Specifications

Melting Point 45°C
Boiling Point 219°C
Quantity 5 g
Formula Weight 95.10
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Pyrrole-2-carboxaldehyde